Methyl-1-(4-chlorophenyl)-4-oxo-1,4-dihydro-3-pyridazinecarboxylate 95%

Lieferant: Apollo Scientific

OR33151-1G OR33151-5G
APOSOR33151-1GEA 270 EUR
APOSOR33151-1G APOSOR33151-5G
Methyl-1-(4-chlorophenyl)-4-oxo-1,4-dihydro-3-pyridazinecarboxylate 95%
Methyl-1-(4-chlorophenyl)-4-oxo-1,4-dihydro-3-pyridazinecarboxylate
Formel: C₁₂H₉ClN₂O₃
Molecular Weight: 264,67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03001289
CAS-Nummer: 453557-76-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR