3-(Methylsulphanyl)benzenol 95%
Lieferant: Apollo Scientific
Synonyme:
3-(Methylsulphanyl)phenol
OR33162-1G
OR33162-5G
APOSOR33162-1GEA
270
EUR
APOSOR33162-1G
APOSOR33162-5G
3-(Methylsulphanyl)benzenol 95%
3-(Methylsulphanyl)benzenol
Formel:
C₇H₈OS Molecular Weight: 140.21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11207488 CAS-Nummer: 3463-03-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...