7-Methylimidazo[1,2-a]pyridin-2-carbonsäure 95%

Lieferant: Apollo Scientific

OR33175-5G
APOSOR33175-5GEA 270 EUR
APOSOR33175-5G
7-Methylimidazo[1,2-a]pyridin-2-carbonsäure 95%
7-Methylimidazo[1,2-a]pyridin-2-carbonsäure
Formel: C₉H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03425067
CAS-Nummer: 80353-94-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR