4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzeneboronic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR3319-1G
OR3319-5G
APOSOR3319-1GEA
299
EUR
APOSOR3319-1G
APOSOR3319-5G
4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzeneboronic acid 98%
4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzeneboronic acid
Formel:
C₉H₉BN₂O₃ Schmelzpunkt: 188…190 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08436062 CAS-Nummer: 913835-70-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...