3-(Dimethylamino)-1-(3-thienyl)-2-buten-1-one 95%

Lieferant: Apollo Scientific

OR33195-1G OR33195-5G
APOSOR33195-1GEA 336 EUR
APOSOR33195-1G APOSOR33195-5G
3-(Dimethylamino)-1-(3-thienyl)-2-buten-1-one 95%
3-(Dimethylamino)-1-(3-thienyl)-2-buten-1-one
Formel: C₁₀H₁₃NOS
Molecular Weight: 195,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659591
CAS-Nummer: 885949-97-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR