3-Acetamido-5-carboxyphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Acetamido-5-boronobenzoic acid
OR3320-1G
OR3320-5G
APOSOR3320-1GEA
318
EUR
APOSOR3320-1G
APOSOR3320-5G
3-Acetamido-5-carboxyphenylboronsäure 98%
3-Acetamido-5-carboxyphenylboronsäure
Formel:
C₉H₁₀BNO₅ Schmelzpunkt: 240…243 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08436066 CAS-Nummer: 108749-15-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...