4-Methyl-3-(1H-pyrrol-1-yl)aniline 95%

Lieferant: Apollo Scientific

OR33204-1G OR33204-5G
APOSOR33204-1GEA 538 EUR
APOSOR33204-1G APOSOR33204-5G
4-Methyl-3-(1H-pyrrol-1-yl)aniline 95%
4-Methyl-3-(1H-pyrrol-1-yl)aniline
Formel: C₁₁H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07643299
CAS-Nummer: 94009-17-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR