5-Chloro-1-(4-chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid 95%

Lieferant: Apollo Scientific

OR33217-1G OR33217-5G
APOSOR33217-1GEA 306 EUR
APOSOR33217-1G APOSOR33217-5G
5-Chloro-1-(4-chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid 95%
5-Chloro-1-(4-chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid
Formel: C₁₃H₉Cl₂NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00140420
CAS-Nummer: 339024-38-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR