5-Chloro-1-(3-chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid 95%
Lieferant: Apollo Scientific
OR33218-1G
OR33218-5G
APOSOR33218-1GEA
306
EUR
APOSOR33218-1G
APOSOR33218-5G
5-Chloro-1-(3-chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid 95%
5-Chloro-1-(3-chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid
Formel:
C₁₃H₉Cl₂NO₃ Molecular Weight: 298.13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00140433 CAS-Nummer: 339024-74-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...