N-(2-Hydroxyethyl)-2-phenylacetamid 95%

Lieferant: Apollo Scientific

OR33222-1G OR33222-25G OR33222-5G
APOSOR33222-1GEA 180 EUR
APOSOR33222-1G APOSOR33222-25G APOSOR33222-5G
N-(2-Hydroxyethyl)-2-phenylacetamid 95%
N-(2-Hydroxyethyl)-2-phenylacetamid
Formel: C₁₀H₁₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00043506
CAS-Nummer: 6269-99-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR