2-(Aminooxy)-1-(4-phenylpiperazino)-1-ethanone hydrochloride 95%

Lieferant: Apollo Scientific

OR33230-1G OR33230-5G
APOSOR33230-1GEA 336 EUR
APOSOR33230-1G APOSOR33230-5G
2-(Aminooxy)-1-(4-phenylpiperazino)-1-ethanone hydrochloride 95%
2-(Aminooxy)-1-(4-phenylpiperazino)-1-ethanone hydrochloride
Formel: C₁₂H₁₈ClN₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01114500
CAS-Nummer: 275374-83-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR