5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carbonitrile 95%

Lieferant: Apollo Scientific

OR33239-1G OR33239-5G
APOSOR33239-1GEA 306 EUR
APOSOR33239-1G APOSOR33239-5G
5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carbonitrile 95%
5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carbonitrile
Formel: C₁₁H₈ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01935950
CAS-Nummer: 321848-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR