Methyl-2-[(3-chloro-2,2-dimethylpropanoyl)amino]benzenecarboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-(3-chloro-2,2-dimethylpropanamido)benzoate

OR33243-1G OR33243-25G OR33243-5G
APOSOR33243-1GEA 180 EUR
APOSOR33243-1G APOSOR33243-25G APOSOR33243-5G
Methyl-2-[(3-chloro-2,2-dimethylpropanoyl)amino]benzenecarboxylate 95%
Methyl-2-[(3-chloro-2,2-dimethylpropanoyl)amino]benzenecarboxylate
Formel: C₁₃H₁₆ClNO₃
Molecular Weight: 269.73 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793430
CAS-Nummer: 339100-86-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR