4-Benzooxazol-2-ylphenylamine 95%

Lieferant: Apollo Scientific

Synonyme: 4-(1,3-Benzoxazol-2-yl)aniline

OR33261-5G
APOSOR33261-5GEA 74.1 EUR
APOSOR33261-5G
4-Benzooxazol-2-ylphenylamine 95%
4-Benzooxazol-2-ylphenylamine
Formel: C₁₃H₁₀N₂O
Molecular Weight: 210.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00453046
CAS-Nummer: 20934-81-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR