3-(1-Azepanyl)-1-propanamine 95%
Lieferant: Apollo Scientific
Synonyme:
3-(Azepan-1-yl)propan-1-amine
OR33267-1G
OR33267-5G
APOSOR33267-1GEA
180
EUR
APOSOR33267-1G
APOSOR33267-5G
3-(1-Azepanyl)-1-propanamine 95%
3-(1-Azepanyl)-1-propanamine
Formel:
C₉H₂₀N₂ Molecular Weight: 156.27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00020990 CAS-Nummer: 3437-33-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...