5-Bromo-2-(1H-pyrrol-1-yl)pyrimidine 95%
Lieferant: Apollo Scientific
OR33271-1G
OR33271-5G
APOSOR33271-1GEA
163
EUR
APOSOR33271-1G
APOSOR33271-5G
5-Bromo-2-(1H-pyrrol-1-yl)pyrimidine 95%
5-Bromo-2-(1H-pyrrol-1-yl)pyrimidine
Formel:
C₈H₆BrN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01871728 CAS-Nummer: 478258-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...