4-Amino-3,5-diphenyl-1,2,4-triazol 95%

Lieferant: Apollo Scientific

Synonyme: Diphenyl-4H-1,2,4-triazol-4-amine, 3,5-Diphenyl-4H-1,2,4-triazol-4-amine

OR33274-1G
APOSOR33274-1GEA 112 EUR
APOSOR33274-1G
4-Amino-3,5-diphenyl-1,2,4-triazol 95%
4-Amino-3,5-diphenyl-1,2,4-triazole
Formel: C₁₄H₁₂N₄
Molecular Weight: 236,2700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00450576
CAS-Nummer: 3049-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR