Ethyl-4-(3-formyl-2,5-dimethyl-1H-pyrrol-1-yl)benzenecarboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 4-(3-formyl-2,5-dimethyl-1H-pyrrol-1-yl)benzoate
OR33275-25G
OR33275-5G
APOSOR33275-25GEA
592
EUR
APOSOR33275-25G
APOSOR33275-5G
Ethyl-4-(3-formyl-2,5-dimethyl-1H-pyrrol-1-yl)benzenecarboxylate 95%
Ethyl-4-(3-formyl-2,5-dimethyl-1H-pyrrol-1-yl)benzenecarboxylate
Formel:
C₁₆H₁₇NO₃ Molecular Weight: 271,32 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022461 CAS-Nummer: 52034-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...