Ethyl-1-(2-cyanophenyl)-4-piperidinecarboxylate 95%
Lieferant: Apollo Scientific
OR33289-1G
OR33289-5G
OR33289-25G
APOSOR33289-1GEA
180
EUR
APOSOR33289-1G
APOSOR33289-5G
APOSOR33289-25G
Ethyl-1-(2-cyanophenyl)-4-piperidinecarboxylate 95%
Ethyl-1-(2-cyanophenyl)-4-piperidinecarboxylate
Formel:
C₁₅H₁₈N₂O₂ Molecular Weight: 258,32 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04117820 CAS-Nummer: 357670-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...