2-Amino-N-(2-propynyl)benzenecarboxamide 95%
Lieferant: Apollo Scientific
Synonyme:
2-Amino-N-(prop-2-yn-1-yl)benzamide
OR33291-1G
OR33291-5G
APOSOR33291-1GEA
163
EUR
APOSOR33291-1G
APOSOR33291-5G
2-Amino-N-(2-propynyl)benzenecarboxamide 95%
2-Amino-N-(2-propynyl)benzenecarboxamide
Formel:
C₁₀H₁₀N₂O Molecular Weight: 174,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425820 CAS-Nummer: 4943-83-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...