6-(4-Benzylpiperazino)nicotinic acid 95%
Lieferant: Apollo Scientific
Synonyme:
6-(4-Benzylpiperazin-1-yl)pyridine-3-carboxylic acid
OR33294-1G
OR33294-5G
APOSOR33294-1GEA
306
EUR
APOSOR33294-1G
APOSOR33294-5G
6-(4-Benzylpiperazino)nicotinic acid 95%
6-(4-Benzylpiperazino)nicotinic acid
Formel:
C₁₇H₁₉N₃O₂ Molecular Weight: 297,36 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03791243 CAS-Nummer: 886360-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...