4-Chloro[1,2,4]triazolo[4,3-a]quinoxaline 95%
Lieferant: Apollo Scientific
Synonyme:
4-Chloro-[1,2,4]triazolo[4,3-a]quinoxaline
OR33340-1G
APOSOR33340-1GEA
336
EUR
APOSOR33340-1G
4-Chloro[1,2,4]triazolo[4,3-a]quinoxaline 95%
4-Chloro[1,2,4]triazolo[4,3-a]quinoxaline
Formel:
C₉H₅ClN₄ Molecular Weight: 204,62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00140798 CAS-Nummer: 62603-54-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...