3-Chlor-2-(2-bromacetyl)thiophen 95%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-1-(3-chlorothiophen-2-yl)ethan-1-one

OR33359-1G OR33359-5G
APOSOR33359-1GEA 538 EUR
APOSOR33359-1G APOSOR33359-5G
3-Chlor-2-(2-bromacetyl)thiophen 95%
3-Chlor-2-(2-bromacetyl)thiophen
Formel: C₆H₄BrClOS
Lagertemperatur: Raumtemperatur
MDL: MFCD00793892
CAS-Nummer: 256486-71-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR