4-[3-(2-Furyl)-1H-pyrazol-5-yl]piperidine 95%

Lieferant: Apollo Scientific

OR33362-1G OR33362-5G
APOSOR33362-1GEA 270 EUR
APOSOR33362-1G APOSOR33362-5G
4-[3-(2-Furyl)-1H-pyrazol-5-yl]piperidine 95%
4-[3-(2-Furyl)-1H-pyrazol-5-yl]piperidine
Formel: C₁₂H₁₅N₃O
Molecular Weight: 217,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00665128
CAS-Nummer: 111897-11-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR