3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydro-2-naphthalinboronsäure 95%
Lieferant: Apollo Scientific
Synonyme:
(3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)boronic acid
OR3337-1G
OR3337-250MG
APOSOR3337-1GEA
369
EUR
APOSOR3337-1G
APOSOR3337-250MG
3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydro-2-naphthalinboronsäure 95%
3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydro-2-naphthalinboronsäure
Formel:
C₁₅H₂₃BO₂ Molecular Weight: 246.1529 g/mol Schmelzpunkt: 198…201 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08436043 CAS-Nummer: 169126-64-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...