2-Chloro-N-cyclohexylnicotinamide 95%
Lieferant: Apollo Scientific
Synonyme:
2-Chloro-N-cyclohexylpyridine-3-carboxamide
OR33386-1G
OR33386-5G
APOSOR33386-1GEA
224
EUR
APOSOR33386-1G
APOSOR33386-5G
2-Chloro-N-cyclohexylnicotinamide 95%
2-Chloro-N-cyclohexylnicotinamide
Formel:
C₁₂H₁₅ClN₂O Molecular Weight: 238,72 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01344059 CAS-Nummer: 57841-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...