3-(4-Methylphenyl)-3-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid 95%

Lieferant: Apollo Scientific

OR33400-1G OR33400-5G
APOSOR33400-1GEA 336 EUR
APOSOR33400-1G APOSOR33400-5G
3-(4-Methylphenyl)-3-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid 95%
3-(4-Methylphenyl)-3-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid
Formel: C₁₈H₁₇NO₃
Molecular Weight: 295.34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425823
CAS-Nummer: 439095-70-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR