4'-Brom-2'-hydroxypropiophenon 95%

Lieferant: Apollo Scientific

Synonyme: 1-(4-Bromo-2-hydroxyphenyl)propan-1-one

OR33409-1G
APOSOR33409-1GEA 180 EUR
APOSOR33409-1G
4'-Brom-2'-hydroxypropiophenon 95%
4'-Brom-2'-hydroxypropiophenon
Formel: C₉H₉BrO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03791251
CAS-Nummer: 17764-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR