2-Amino-6-methyl-3-nitropyridin 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Methyl-3-nitropyridin-2-amin, 2-Amino-3-nitro-6-picolin, 6-Methyl-3-nitro-2-pyridylamin

OR3342-1G OR3342-5G
APOSOR3342-1GEA 21.9 EUR
APOSOR3342-1G APOSOR3342-5G
2-Amino-6-methyl-3-nitropyridin 95%
2-Amino-6-methyl-3-nitropyridin
Formel: C₆H₇N₃O₂
Molecular Weight: 153.14 g/mol
Schmelzpunkt: 147…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00047443
CAS-Nummer: 21901-29-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR