1-[4-(3-Chloropropoxy)-2-hydroxyphenyl]-1-ethanone 95%

Lieferant: Apollo Scientific

OR33421-1G OR33421-25G OR33421-5G
APOSOR33421-1GEA 180 EUR
APOSOR33421-1G APOSOR33421-25G APOSOR33421-5G
1-[4-(3-Chloropropoxy)-2-hydroxyphenyl]-1-ethanone 95%
1-[4-(3-Chloropropoxy)-2-hydroxyphenyl]-1-ethanone
Formel: C₁₁H₁₃ClO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04126369
CAS-Nummer: 172739-45-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR