5-(2-Bromphenyl)oxazol 95%

Lieferant: Apollo Scientific

Synonyme: 5-(2-Bromophenyl)-1,3-oxazole

OR33433-5G
APOSOR33433-5GEA 58 EUR
APOSOR33433-5G
5-(2-Bromphenyl)oxazol 95%
5-(2-Bromophenyl)oxazole
Formel: C₉H₆BrNO
Molecular Weight: 224,0540 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05668985
CAS-Nummer: 328270-70-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR