Ethyl-3-formyl-1H-indole-2-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 3-formyl-1H-indole-2-carboxylate

OR33448-1G OR33448-5G
APOSOR33448-1GEA 180 EUR
APOSOR33448-1G APOSOR33448-5G
Ethyl-3-formyl-1H-indole-2-carboxylate 95%
Ethyl-3-formyl-1H-indole-2-carboxylate
Formel: C₁₂H₁₁NO₃
Molecular Weight: 217.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01463252
CAS-Nummer: 18450-27-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR