4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzenecarbaldehyde 95%
Lieferant: Apollo Scientific
Synonyme:
4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzaldehyde
OR33451-1G
OR33451-5G
APOSOR33451-1GEA
336
EUR
APOSOR33451-1G
APOSOR33451-5G
4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzenecarbaldehyde 95%
4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzenecarbaldehyde
Formel:
C₁₆H₁₄O₃ Molecular Weight: 254.29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09152704 CAS-Nummer: 952182-92-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...