2-Chloro-N-(2-hydroxyethyl)-4-piperidinobenzenecarboxamide 95%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-N-(2-hydroxyethyl)-4-(piperidin-1-yl)benzamide

OR33456-1G OR33456-5G
APOSOR33456-1GEA 336 EUR
APOSOR33456-1G APOSOR33456-5G
2-Chloro-N-(2-hydroxyethyl)-4-piperidinobenzenecarboxamide 95%
2-Chloro-N-(2-hydroxyethyl)-4-piperidinobenzenecarboxamide
Formel: C₁₄H₁₉ClN₂O₂
Molecular Weight: 282,77 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09152713
CAS-Nummer: 952183-00-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR