2-(4-Methylphenyl)-2-oxoethyl acetate 95%
Lieferant: Apollo Scientific
Synonyme:
2-Oxo-2-(p-tolyl)ethyl acetate
OR33457-1G
OR33457-25G
OR33457-5G
APOSOR33457-1GEA
180
EUR
APOSOR33457-1G
APOSOR33457-25G
APOSOR33457-5G
2-(4-Methylphenyl)-2-oxoethyl acetate 95%
2-(4-Methylphenyl)-2-oxoethyl acetate
Formel:
C₁₁H₁₂O₃ Molecular Weight: 192,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00458047 CAS-Nummer: 65143-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...