Methyl-2-chloro-4-(1H-1,2,4-triazol-1-yl)benzenecarboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-chloro-4-(1H-1,2,4-triazol-1-yl)benzoate

OR33458-1G OR33458-5G
APOSOR33458-1GEA 270 EUR
APOSOR33458-1G APOSOR33458-5G
Methyl-2-chloro-4-(1H-1,2,4-triazol-1-yl)benzenecarboxylate 95%
Methyl-2-chloro-4-(1H-1,2,4-triazol-1-yl)benzenecarboxylate
Formel: C₁₀H₈ClN₃O₂
Molecular Weight: 237,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09152714
CAS-Nummer: 220141-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR