2-(4-Benzylpiperidino)acetonitrile 95%

Lieferant: Apollo Scientific

OR33464-1G OR33464-5G
APOSOR33464-1GEA 270 EUR
APOSOR33464-1G APOSOR33464-5G
2-(4-Benzylpiperidino)acetonitrile 95%
2-(4-Benzylpiperidino)acetonitrile
Formel: C₁₄H₁₈N₂
Molecular Weight: 214,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09152769
CAS-Nummer: 25842-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR