3-(2-Methyl-1H-indol-3-yl)-3-oxopropanenitrile 95%

Lieferant: Apollo Scientific

OR33475-1G OR33475-5G
APOSOR33475-1GEA 180 EUR
APOSOR33475-1G APOSOR33475-5G
3-(2-Methyl-1H-indol-3-yl)-3-oxopropanenitrile 95%
3-(2-Methyl-1H-indol-3-yl)-3-oxopropanenitrile
Formel: C₁₂H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD02330570
CAS-Nummer: 76397-72-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR