4-Brom-2-methyl-3-chinolinol 95%
Lieferant: Apollo Scientific
OR33495-1G
APOSOR33495-1GEA
209
EUR
APOSOR33495-1G
4-Brom-2-methyl-3-chinolinol 95%
4-Brom-2-methyl-3-chinolinol
Formel:
C₁₀H₈BrNO Molecular Weight: 238,08 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
13235-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...