2-Bromo-3,6-dihydroxybenzenecarbaldehyde 95%
Lieferant: Apollo Scientific
OR33496-1G
OR33496-5G
APOSOR33496-1GEA
270
EUR
APOSOR33496-1G
APOSOR33496-5G
2-Bromo-3,6-dihydroxybenzenecarbaldehyde 95%
2-Bromo-3,6-dihydroxybenzenecarbaldehyde
Formel:
C₇H₅BrO₃ Molecular Weight: 217,03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14584781 CAS-Nummer: 241127-72-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...