1-Thia-4-azaspiro[4.4]nonan-3-one 95%

Lieferant: Apollo Scientific

OR33501-1G OR33501-5G
APOSOR33501-1GEA 270 EUR
APOSOR33501-1G APOSOR33501-5G
1-Thia-4-azaspiro[4.4]nonan-3-one 95%
1-Thia-4-azaspiro[4.4]nonan-3-one
Formel: C₇H₁₁NOS
Lagertemperatur: Raumtemperatur
MDL: MFCD00101404
CAS-Nummer: 72752-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR