2-Aminoquinolin-4-ol 95%

Lieferant: Apollo Scientific

OR33507-1G OR33507-5G
APOSOR33507-1GEA 88.6 EUR
APOSOR33507-1G APOSOR33507-5G
2-Aminoquinolin-4-ol 95%
2-Aminoquinolin-4-ol
Formel: C₉H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00052386
CAS-Nummer: 42712-64-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR