4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzenecarboxylic acid 95%
Lieferant: Apollo Scientific
Synonyme:
4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzoic acid
OR33514-1G
OR33514-5G
APOSOR33514-1GEA
336
EUR
APOSOR33514-1G
APOSOR33514-5G
4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzenecarboxylic acid 95%
4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzenecarboxylic acid
Formel:
C₁₆H₁₄O₄ Molecular Weight: 270,29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09152731 CAS-Nummer: 952183-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...