2-Amino-6-bromo-4-quinolinol 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR33516-1G
OR33516-5G
APOSOR33516-1GEA
336
EUR
APOSOR33516-1G
APOSOR33516-5G
2-Amino-6-bromo-4-quinolinol 95%
2-Amino-6-bromo-4-quinolinol
Formel:
C₉H₇BrN₂O Molecular Weight: 239,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08234781 CAS-Nummer: 123420-09-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...