5-Chloro-1-(4-chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde 95%

Lieferant: Apollo Scientific

OR33518-1G OR33518-5G
APOSOR33518-1GEA 671 EUR
APOSOR33518-1G APOSOR33518-5G
5-Chloro-1-(4-chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde 95%
5-Chloro-1-(4-chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
Formel: C₁₃H₉Cl₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10758059
CAS-Nummer: 1033463-21-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR