3-Oxo-3-(4-piperidinophenyl)propanenitrile 95%

Lieferant: Apollo Scientific

OR33527-1G OR33527-5G
APOSOR33527-1GEA 336 EUR
APOSOR33527-1G APOSOR33527-5G
3-Oxo-3-(4-piperidinophenyl)propanenitrile 95%
3-Oxo-3-(4-piperidinophenyl)propanenitrile
Formel: C₁₄H₁₆N₂O
Molecular Weight: 228.3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12025857
CAS-Nummer: 1135283-23-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR