3-(5-Bromo-1-benzofuran-2-yl)-3-oxopropanenitrile 95%

Lieferant: Apollo Scientific

OR33528-1G OR33528-5G
APOSOR33528-1GEA 336 EUR
APOSOR33528-1G APOSOR33528-5G
3-(5-Bromo-1-benzofuran-2-yl)-3-oxopropanenitrile 95%
3-(5-Bromo-1-benzofuran-2-yl)-3-oxopropanenitrile
Formel: C₁₁H₆BrNO₂
Molecular Weight: 264.08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12025858
CAS-Nummer: 1135283-25-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR