1-(tert-Butyl)-5-methyl-1H-pyrazole-4-carboxylic acid 95%
Lieferant: Apollo Scientific
OR33553-1G OR33553-5G
APOSOR33553-1GEA 538 EUR
APOSOR33553-1G APOSOR33553-5G
1-(tert-Butyl)-5-methyl-1H-pyrazole-4-carboxylic acid 95%
1-(tert-Butyl)-5-methyl-1H-pyrazole-4-carboxylic acid
Formel: C₉H₁₄N₂O₂
Molecular Weight: 182,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11810587
CAS-Nummer: 288251-51-4