2,4-Dichloro-6,7-dihydro-5H-cyclopenta[d]pyrimidine 95%

Lieferant: Apollo Scientific

OR33554-5G
APOSOR33554-5GEA 111 EUR
APOSOR33554-5G
2,4-Dichloro-6,7-dihydro-5H-cyclopenta[d]pyrimidine 95%
2,4-Dichloro-6,7-dihydro-5H-cyclopenta[d]pyrimidine
Formel: C₇H₆Cl₂N₂
Molecular Weight: 189,0419 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09750204
CAS-Nummer: 5466-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR