2-Chloro-4-methoxy-5H,6H,7H-cyclopenta[d]pyrimidine 95%

Lieferant: Apollo Scientific

OR33556-1G OR33556-100MG OR33556-250MG
APOSOR33556-1GEA 986 EUR
APOSOR33556-1G APOSOR33556-100MG APOSOR33556-250MG
2-Chloro-4-methoxy-5H,6H,7H-cyclopenta[d]pyrimidine 95%
2-Chloro-4-methoxy-5H,6H,7H-cyclopenta[d]pyrimidine
Formel: C₈H₉ClN₂O
Molecular Weight: 184.63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651768
CAS-Nummer: 81532-47-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR